5-(Trifluoromethyl)benzo[2,1,3]thiadiazole 97%
Supplier: Apollo Scientific
Warning
PC3212-1G PC3212-5G
APOSPC3212-1GEA 89.9 EUR
APOSPC3212-1G APOSPC3212-5G
5-(Trifluoromethyl)benzo[2,1,3]thiadiazole 97%
5-(Trifluoromethyl)benzo[2,1,3]thiadiazole
Formula: C₇H₃F₃N₂S
Boiling Pt: 100 °C/15mmHg
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00114584
CAS Number: 17754-05-1