Ethyl-3-fluorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-fluorobenzoate, 3-Fluorobenzoic acid ethyl ester
PC3220-100G PC3220-25G
APOSPC3220-100GEA 46.4 EUR
APOSPC3220-100G APOSPC3220-25G
Ethyl-3-fluorobenzoate 98%
Ethyl 3-fluorobenzoate
Formula: C₉H₉FO₂
Boiling Pt: 94…95 °C/16mmHg
Density: 1.136 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000336
CAS Number: 451-02-5
EINECS: 207-191-1