2-Fluorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Danger
PC32287-25G PC32287-5G
APOSPC32287-25GEA 248 EUR
APOSPC32287-25G APOSPC32287-5G
2-Fluorophenyl isothiocyanate 97%
2-Fluorophenyl isothiocyanate
Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 103…104 °C/12mmHg
Melting Pt: 24…26 °C
Density: 1.248 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004800
CAS Number: 38985-64-7
EINECS: 254-237-1
UN: 2922
ADR: 8+6.1 II