3-Fluoro-4-nitropyridine 95%
Supplier: Apollo Scientific
Warning
PC3238-5G PC3238-1G PC3238-250MG
APOSPC3238-5GEA 458 EUR
APOSPC3238-5G APOSPC3238-1G APOSPC3238-250MG
3-Fluoro-4-nitropyridine 95%
3-Fluoro-4-nitropyridine
Formula: C₅H₃FN₂O₂
Boiling Pt: 62…64 °C/5mmHg
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112517
CAS Number: 13505-01-6
UN: 2810
ADR: 6.1,III