Ethyl 2-methyl-4,4,4-trifluoroacetoacetate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4,4,4-Trifluoro-2-methyl-3-oxobutyrate
PC3245D-25G PC3245D-5G PC3245D-100G
APOSPC3245D-25GEA 116 EUR
APOSPC3245D-25G APOSPC3245D-5G APOSPC3245D-100G
Ethyl 2-methyl-4,4,4-trifluoroacetoacetate 97%
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate
Formula: C₇H₉F₃O₃
Boiling Pt: 144…146 °C
Density: 1.258 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00190635
CAS Number: 344-00-3
UN: 1993
ADR: 3,III