1-[6-(4-Fluorophenyl)pyridin-3-yl]ethanone
Supplier: Apollo Scientific
PC32876-1G PC32876-100MG PC32876-250MG PC32876-500MG PC32876-2.5G
APOSPC32876-1GEA 580 EUR
APOSPC32876-1G APOSPC32876-100MG APOSPC32876-250MG APOSPC32876-500MG APOSPC32876-2.5G
1-[6-(4-Fluorophenyl)pyridin-3-yl]ethanone
1-[6-(4-Fluorophenyl)pyridin-3-yl]ethanone
Formula: C₁₃H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD12404910
CAS Number: 1215695-69-4