Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl (2H-perfluoroprop-2-yl)acetate
PC3289E-5G PC3289E-25G PC3289E-1G
APOSPC3289E-5GEA 139 EUR
APOSPC3289E-5G APOSPC3289E-25G APOSPC3289E-1G
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 97%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Formula: C₇H₈F₆O₂
MW: 238.13 g/mol
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00236608
CAS Number: 17327-34-3
UN: 3272
ADR: 3 II