2-Chloro-3-fluorobenzoic acid
Supplier: Apollo Scientific
Warning
PC3299-25G PC3299-5G PC3299-1G
APOSPC3299-25GEA 58 EUR
APOSPC3299-25G APOSPC3299-5G APOSPC3299-1G
2-Chloro-3-fluorobenzoic acid
2-Chloro-3-fluorobenzoic acid
Formula: C₇H₄ClFO₂
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00973903
CAS Number: 102940-86-3