2,2,2-Trifluoroethyl crotonate 99%
Supplier: Apollo Scientific
Warning
PC3394-25G PC3394-5G PC3394-1G
APOSPC3394-25GEA 272 EUR
APOSPC3394-25G APOSPC3394-5G APOSPC3394-1G
2,2,2-Trifluoroethyl crotonate 99%
2,2,2-Trifluoroethyl crotonate
Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 52…53 °C/115mmHg
Density: 1.18 g/cm³ (25 °C)
Flash Pt: <37 °C
Storage Temperature: Ambient
MDL Number: MFCD04038364
CAS Number: 287957-36-2
UN: 1993
ADR: 3 III