2-Fluorobenzamide 98%
Supplier: Apollo Scientific
Warning
PC3472-5G PC3472-25G
APOSPC3472-5GEA 56.7 EUR
APOSPC3472-5G APOSPC3472-25G
2-Fluorobenzamide 98%
2-Fluorobenzamide
Formula: C₇H₆FNO
MW: 139.13 g/mol
Boiling Pt: 287
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00007970
CAS Number: 445-28-3
EINECS: 207-157-6