2,4,5-Trifluoro-4-hydroxybenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC3496-25G PC3496-100G
APOSPC3496-25GEA 97.3 EUR
APOSPC3496-25G APOSPC3496-100G
2,4,5-Trifluoro-4-hydroxybenzoic acid 98%
2,4,5-Trifluoro-4-hydroxybenzoic acid
Formula: C₇H₃F₃O₃
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00800378
CAS Number: 116751-24-7