2',6'-Dichloro-3'-fluoroacetophenone 98%
Supplier: Apollo Scientific
Warning
PC3497-100G PC3497-25G
APOSPC3497-100GEA 27.7 EUR
APOSPC3497-100G APOSPC3497-25G
2',6'-Dichloro-3'-fluoroacetophenone 98%
2',6'-Dichloro-3'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
Boiling Pt: 250…255 °C
Density: 1.403 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD02093760
CAS Number: 290835-85-7