2-Fluorobenzoic acid 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
o-Fluorobenzoic acid
PC3500-1KG
PC3500-100G
PC3500-500G
APOSPC3500-1KGEA
55.1
EUR
APOSPC3500-1KG
APOSPC3500-100G
APOSPC3500-500G
2-Fluorobenzoic acid 99%
2-Fluorobenzoic acid
Formula:
C₇H₅FO₂ MW: 140.11 g/mol Boiling Pt: 245 Melting Pt: 122…125 °C Density: 1.46 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |