2-Fluorobenzoyl chloride 99%
Supplier: Apollo Scientific
Danger
PC3610-100G PC3610-500G
APOSPC3610-100GEA 67.3 EUR
APOSPC3610-100G APOSPC3610-500G
2-Fluorobenzoyl chloride 99%
2-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
Boiling Pt: 90…92 °C/15mmHg
Melting Pt: 44351 °C
Density: 1.328 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II