2-Fluoroethylamine hydrochloride 98%
Supplier: Apollo Scientific
Danger
PC3677-1G PC3677-25G PC3677-5G
APOSPC3677-1GEA 66.7 EUR
APOSPC3677-1G APOSPC3677-25G APOSPC3677-5G
2-Fluoroethylamine hydrochloride 98%
2-Fluoroethylamine hydrochloride
Formula: C₂H₆FN·HCl
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00008161
CAS Number: 460-08-2
EINECS: 207-302-3
UN: 2811
ADR: 6.1,II