α-Bromo-4-fluorotoulene 98%
Supplier: Apollo Scientific
Danger
PC3695-100G PC3695-25G PC3695-500G
APOSPC3695-100GEA 48 EUR
APOSPC3695-100G APOSPC3695-25G APOSPC3695-500G
α-Bromo-4-fluorotoulene 98%
α-Bromo-4-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 85 °C/15mmHg
Density: 1.517 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000359
CAS Number: 459-46-1
UN: 3265
ADR: 8,II