α-Chloro-3-fluorotoulene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride
PC3710-100G PC3710-1KG PC3710-500G
APOSPC3710-100GEA 74.1 EUR
APOSPC3710-100G APOSPC3710-1KG APOSPC3710-500G
α-Chloro-3-fluorotoulene 97%
α-Chloro-3-fluorotoulene
Formula: C₇H₆ClF
Boiling Pt: 84 °C/29mmHg
Density: 1.213 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00000904
CAS Number: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8+3,II