Benzyl-2,2,2-trifluoroethyl sulphide 97%
Supplier: Apollo Scientific
Danger
PC3722-25G PC3722-5G
APOSPC3722-25GEA 395 EUR
APOSPC3722-25G APOSPC3722-5G
Benzyl-2,2,2-trifluoroethyl sulphide 97%
Benzyl-2,2,2-trifluoroethyl sulphide
Formula: C₉H₉F₃S
Boiling Pt: 45…46 °C/20mmHg
Density: 1.222 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00191786
CAS Number: 77745-03-0
UN: 1993
ADR: 3,II