1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether 97%
Supplier: Apollo Scientific
Warning
PC3813-1G PC3813-25G PC3813-5G
APOSPC3813-1GEA 46.4 EUR
APOSPC3813-1G APOSPC3813-25G APOSPC3813-5G
1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether 97%
1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether
Formula: C₅H₃F₉O
Boiling Pt: 72.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00476468
CAS Number: 993-95-3