2-Fluoro-6-methoxyphenol 98%
Supplier: Apollo Scientific
Warning
PC3817J-25G PC3817J-5G
APOSPC3817J-25GEA 368 EUR
APOSPC3817J-25G APOSPC3817J-5G
2-Fluoro-6-methoxyphenol 98%
2-Fluoro-6-methoxyphenol
Formula: C₇H₇FO₂
Boiling Pt: 130…131 °C/36mmHg
Density: 1.23 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00075140
CAS Number: 73943-41-6