4-Fluoro-3-methylanisole 97%
Supplier: Apollo Scientific
Warning
PC3822D-100G PC3822D-10G PC3822D-25G PC3822D-1G PC3822D-5G
APOSPC3822D-100GEA 0 EUR
APOSPC3822D-100G APOSPC3822D-10G APOSPC3822D-25G APOSPC3822D-1G APOSPC3822D-5G
4-Fluoro-3-methylanisole 97%
4-Fluoro-3-methylanisole
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 174…175 °C/762mmHg
Density: 1.07 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00042293
CAS Number: 2338-54-7