2-(2,3,6-Trifluorophenoxy)ethanol 95+%
Supplier: Apollo Scientific
Warning
PC3843-1G PC3843-5G
APOSPC3843-1GEA 92.8 EUR
APOSPC3843-1G APOSPC3843-5G
2-(2,3,6-Trifluorophenoxy)ethanol 95+%
2-(2,3,6-Trifluorophenoxy)ethanol
Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Boiling Pt: 95…96 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00134279
CAS Number: 72912-49-3