4-Fluoro-2-nitroaniline 97%
Supplier: Apollo Scientific
Warning
PC3870-100G PC3870-25G PC3870-500G
APOSPC3870-100GEA 39.3 EUR
APOSPC3870-100G APOSPC3870-25G APOSPC3870-500G
4-Fluoro-2-nitroaniline 97%
4-Fluoro-2-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 90…94 °C
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0