2-Chloro-4,5-difluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC3923-1G PC3923-25G PC3923-5G
APOSPC3923-1GEA 31.9 EUR
APOSPC3923-1G APOSPC3923-25G APOSPC3923-5G
2-Chloro-4,5-difluorobenzenesulphonyl chloride 97%
2-Chloro-4,5-difluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₂F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD02091388
CAS Number: 67475-58-5
UN: 1760
ADR: 8,III