4-Fluoro-4'-nitrobenzophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Fluorophenyl)(4-nitrophenyl)methanone
PC3938E-1G PC3938E-250MG
APOSPC3938E-1GEA 21.9 EUR
APOSPC3938E-1G APOSPC3938E-250MG
4-Fluoro-4'-nitrobenzophenone 98%
4-Fluoro-4'-nitrobenzophenone
Formula: C₁₃H₈FNO₃
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00053428
CAS Number: 2195-47-3