2-Fluoro-5-nitrobenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene
PC3940-100G PC3940-25G
APOSPC3940-100GEA 55.1 EUR
APOSPC3940-100G APOSPC3940-25G
2-Fluoro-5-nitrobenzotrifluoride 98%
2-Fluoro-5-nitrobenzotrifluoride
Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 105…110 °C/25mmHg
Density: 1.522 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00024576
CAS Number: 400-74-8
EINECS: 206-925-8