5-Fluoro-2-nitrotoluene 98%
Supplier: Apollo Scientific
|
Warning
|
PC4030-100G
PC4030-250G
PC4030-25G
PC4030-500G
APOSPC4030-100GEA
26.1
EUR
APOSPC4030-100G
APOSPC4030-250G
APOSPC4030-25G
APOSPC4030-500G
5-Fluoro-2-nitrotoluene 98%
5-Fluoro-2-nitrotoluene
Formula:
C₇H₆FNO₂ Boiling Pt: 97…98 °C/10mmHg Density: 1.272 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007283 CAS Number: 446-33-3 EINECS: 207-164-4 |