4-Fluoro-3-nitrotoluene 99%
Supplier: Apollo Scientific
Warning
PC4045-100G PC4045-25G PC4045-500G
APOSPC4045-100GEA 46.4 EUR
APOSPC4045-100G APOSPC4045-25G APOSPC4045-500G
4-Fluoro-3-nitrotoluene 99%
4-Fluoro-3-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 241 °C
Melting Pt: 22…30 °C
Density: 1.262 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007060
CAS Number: 446-11-7