N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
Supplier: Apollo Scientific
Warning
Synonyms: N,N-Bis(trifluoromethylsulphonyl)-2-pyridylamine, 2-Pyridyltriflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]pyridine, 1,1,1-Trifluoro-N-(pyridin-2-yl)-N-((trifluoromethyl)sulphonyl)methanesulphonamide
PC4064-5G PC4064-10G PC4064-1G
APOSPC4064-5GEA 33.5 EUR
APOSPC4064-5G APOSPC4064-10G APOSPC4064-1G
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
N-(2-Pyridyl)bis(trifluoromethanesulphonimide)
Triflating reagent.
Formula: C₇H₄F₆N₂O₄S₂
MW: 358.23 g/mol
Boiling Pt: 80…90 °C/0.25mmHg
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191834
CAS Number: 145100-50-1