2,3-Difluoro-5-nitropyridine 98%
Supplier: Apollo Scientific
PC408056-250MG PC408056-1G
APOSPC408056-250MGEA 98.9 EUR
APOSPC408056-250MG APOSPC408056-1G
2,3-Difluoro-5-nitropyridine 98%
2,3-Difluoro-5-nitropyridine
Formula: C₅H₂F₂N₂O₂
MW: 160.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750303
CAS Number: 954219-68-2