5,5,6,6,7,7,8,8,8-Nonafluorooctan-1-ol
Supplier: Apollo Scientific
Warning
PC408149-1G PC408149-25G PC408149-5G
APOSPC408149-1GEA 198 EUR
APOSPC408149-1G APOSPC408149-25G APOSPC408149-5G
5,5,6,6,7,7,8,8,8-Nonafluorooctan-1-ol
5,5,6,6,7,7,8,8,8-Nonafluorooctan-1-ol
Formula: C₈H₉F₉O
Boiling Pt: 90 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06248594
CAS Number: 3792-02-7
UN: 1987
ADR: 3,III