2-Bromo-6-fluorophenylacetonitrile
Supplier: Apollo Scientific
Danger
PC408235-1G PC408235-250MG
APOSPC408235-1GEA 232 EUR
APOSPC408235-1G APOSPC408235-250MG
2-Bromo-6-fluorophenylacetonitrile
2-Bromo-6-fluorophenylacetonitrile
Formula: C₈H₅BrFN
MW: 214.04 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD11226884
CAS Number: 936693-22-0
UN: 3439
ADR: 6.1 III