8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine 95+%
Supplier: Apollo Scientific
Warning
PC409503-1G PC409503-10G PC409503-5G
APOSPC409503-1GEA 484 EUR
APOSPC409503-1G APOSPC409503-10G APOSPC409503-5G
8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine 95+%
8-Bromo-5-chloro-2-(trifluoromethyl)-1,6-naphthyridine
Formula: C₉H₃BrClF₃N₂
MW: 311.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08567942
CAS Number: 890301-89-0