5-Iodo-2-(trifluoromethyl)pyridine 95+%
Supplier: Apollo Scientific
Warning
PC409520-1G PC409520-5G
APOSPC409520-1GEA 104 EUR
APOSPC409520-1G APOSPC409520-5G
5-Iodo-2-(trifluoromethyl)pyridine 95+%
5-Iodo-2-(trifluoromethyl)pyridine
Formula: C₆H₃F₃IN
MW: 273 g/mol
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD11226772
CAS Number: 873107-98-3