4,4'-Difluorobenzhydryl chloride 97%
Supplier: Apollo Scientific
Danger
PC4106-10G PC4106-50G PC4106-5G
APOSPC4106-10GEA 50.9 EUR
APOSPC4106-10G APOSPC4106-50G APOSPC4106-5G
4,4'-Difluorobenzhydryl chloride 97%
4,4'-Difluorobenzhydryl chloride
Formula: C₁₃H₉ClF₂
Boiling Pt: 126…128 °C/1mmHg
Density: 1.28 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00044329
CAS Number: 27064-94-4
EINECS: 248-201-4
UN: 3265
ADR: 8,II