Heptafluorobutyrylamidine
Supplier: Apollo Scientific
|
Warning
|
PC4125-10G
PC4125-1G
PC4125-5G
APOSPC4125-10GEA
236
EUR
APOSPC4125-10G
APOSPC4125-1G
APOSPC4125-5G
Heptafluorobutyrylamidine
Heptafluorobutyrylamidine
Formula:
C₄H₃F₇N₂ Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236736 CAS Number: 375-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...