1-(4-Fluorophenyl)-2-methyl-2-propylamine 97%
Supplier: Apollo Scientific
Danger
PC4170M-5G PC4170M-25G
APOSPC4170M-5GEA 120 EUR
APOSPC4170M-5G APOSPC4170M-25G
1-(4-Fluorophenyl)-2-methyl-2-propylamine 97%
1-(4-Fluorophenyl)-2-methyl-2-propylamine
Formula: C₁₀H₁₄FN
MW: 167.227 g/mol
Boiling Pt: 180 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00082702
CAS Number: 1200-27-7
UN: 2735
ADR: 8 III