N-tert-Butyl-4-fluoro-2-nitroaniline
Supplier: Apollo Scientific
Warning
PC4355-1G PC4355-250MG PC4355-5G
APOSPC4355-1GEA 139 EUR
APOSPC4355-1G APOSPC4355-250MG APOSPC4355-5G
N-tert-Butyl-4-fluoro-2-nitroaniline
N-tert-Butyl-4-fluoro-2-nitroaniline
Formula: C₁₀H₁₃FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00798108
CAS Number: 239080-03-6