3-Fluoro-5-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Danger
PC4373I-25G PC4373I-5G PC4373I-100G
APOSPC4373I-25GEA 43.5 EUR
APOSPC4373I-25G APOSPC4373I-5G APOSPC4373I-100G
3-Fluoro-5-(trifluoromethyl)benzonitrile 97%
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 212
Melting Pt: 28
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: 6.1 III