4-Fluoro-2-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Cyano-5-fluorobenzotrifluoride
PC4373J-100G PC4373J-25G PC4373J-500G
APOSPC4373J-100GEA 68.3 EUR
APOSPC4373J-100G APOSPC4373J-25G APOSPC4373J-500G
4-Fluoro-2-(trifluoromethyl)benzonitrile 97%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 194…196 °C
Melting Pt: 43…45 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1 III