2-Fluoro-3-(trifluoromethyl)benzophenone 97%
Supplier: Apollo Scientific
Warning
PC4374E-1G PC4374E-5G PC4374E-25G
APOSPC4374E-1GEA 21.9 EUR
APOSPC4374E-1G APOSPC4374E-5G APOSPC4374E-25G
2-Fluoro-3-(trifluoromethyl)benzophenone 97%
2-Fluoro-3-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
MW: 268.21 g/mol
Boiling Pt: 290 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061151
CAS Number: 207853-70-1