2-Fluoro-4-(trifluoromethyl)benzophenone 97%
Supplier: Apollo Scientific
Warning
PC4374F-1G PC4374F-25G PC4374F-5G
APOSPC4374F-1GEA 21.9 EUR
APOSPC4374F-1G APOSPC4374F-25G APOSPC4374F-5G
2-Fluoro-4-(trifluoromethyl)benzophenone 97%
2-Fluoro-4-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
MW: 268.21 g/mol
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00061253
CAS Number: 207974-08-1