2-Fluoro-3-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-3-hydroxybenzotrifluoride
PC4376M-1G PC4376M-25G PC4376M-5G
APOSPC4376M-1GEA 55.1 EUR
APOSPC4376M-1G APOSPC4376M-25G APOSPC4376M-5G
2-Fluoro-3-(trifluoromethyl)phenol 98%
2-Fluoro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 122 °C
Density: 1.431 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00083554
CAS Number: 207291-85-8
UN: 2810
ADR: 6.1 III