Perfluoro(methylcyclohexane)
Supplier: Apollo Scientific
Warning
Synonyms: FlutecR PP2
PC4381N-1KG PC4381N-250G
APOSPC4381N-1KGEA 0 EUR
APOSPC4381N-1KG APOSPC4381N-250G
Perfluoro(methylcyclohexane)
Perfluoro(methylcyclohexane)
Formula: C₇F₁₄
MW: 350.05 g/mol
Boiling Pt: 76 °C
Density: 1.788 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001468
CAS Number: 355-02-2
EINECS: 206-573-5