1H,1H-Heptafluoro-n-butyl methacrylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyl Methacrylate, 1H,1h-heptafluorobutylmethacrylate, Methacrylic acid 2,2,3,3,4,4,4-Heptafluorobutyl ester
PC4412-25G PC4412-5G PC4412-100G
APOSPC4412-25GEA 224 EUR
APOSPC4412-25G APOSPC4412-5G APOSPC4412-100G
1H,1H-Heptafluoro-n-butyl methacrylate 97%
1H,1H-Heptafluoro-n-butyl methacrylate
Formula: C₈H₇F₇O₂
MW: 268.13 g/mol
Boiling Pt: 134…136 °C
Density: 1.345 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00039251
CAS Number: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3 III