5-Chloro-3-(4-fluorophenyl)[1,2,4]thiadiazole
Supplier: Apollo Scientific
Warning
PC446032-1G PC446032-5G
APOSPC446032-1GEA 244 EUR
APOSPC446032-1G APOSPC446032-5G
5-Chloro-3-(4-fluorophenyl)[1,2,4]thiadiazole
5-Chloro-3-(4-fluorophenyl)[1,2,4]thiadiazole
Formula: C₈H₄ClFN₂S
MW: 214.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052447
CAS Number: 138426-27-4