1-Iodoperfluoropropane 95%
Supplier: Apollo Scientific
Warning
Synonyms: Perfluoropropyl iodide, Heptafluoro-1-iodopropane, 1,1,2,2,3,3,3-Heptafluoro-1-iodopropane
PC4510-100G PC4510-25G PC4510-5G
APOSPC4510-100GEA 812 EUR
APOSPC4510-100G APOSPC4510-25G APOSPC4510-5G
1-Iodoperfluoropropane 95%
1-Iodoperfluoropropane
Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 40…41 °C
Melting Pt: –95 °C
Density: 2.05 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001061
CAS Number: 754-34-7
EINECS: 212-045-5
UN: 3163
ADR: 2.2