1H,1H,9H-Hexadecafluorononyl acrylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H,9H-Perfluorononyl acrylate
PC4534-25G PC4534-5G
APOSPC4534-25GEA 499 EUR
APOSPC4534-25G APOSPC4534-5G
1H,1H,9H-Hexadecafluorononyl acrylate 95%
1H,1H,9H-Hexadecafluorononyl acrylate
Formula: C₁₂H₆F₁₆O₂
MW: 486.15 g/mol
Boiling Pt: 75 °C/1mmHg
Density: 1.646 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078281
CAS Number: 4180-26-1
EINECS: 224-053-6