2-Fluoro-3-(trifluoromethyl)benzylamine 98%
Supplier: Apollo Scientific
Danger
PC4538-10G PC4538-5G PC4538-1G
APOSPC4538-10GEA 292 EUR
APOSPC4538-10G APOSPC4538-5G APOSPC4538-1G
2-Fluoro-3-(trifluoromethyl)benzylamine 98%
2-Fluoro-3-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
MW: 193.14 g/mol
Boiling Pt: 193 °C
Density: 1.321 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00061161
CAS Number: 239135-49-0
UN: 2735
ADR: 8 II