1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane
Supplier: Apollo Scientific
Warning
PC4581-1G PC4581-25G PC4581-5G
APOSPC4581-1GEA 33.5 EUR
APOSPC4581-1G APOSPC4581-25G APOSPC4581-5G
1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane
1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane
Formula: F₂CCFOCF₂CF(CF₃)O(CF₂)₂CF₃
Boiling Pt: 103…104 °C
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00191479
CAS Number: 1644-11-7